Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pisy-1
Entrez Gene ID
174902
Gene Name
CDP-diacylglycerol--inositol 3-phosphatidyltransferase
Gene Aliases
CELE_Y46G5A.5
Chromosome Location
Chr.II: 12737032 - 12738338 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_064310.4 NP_496711.1 1 - 2 151 66

View Full Product Details