Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cpt-1
Entrez Gene ID
174912
Gene Name
Carnitine Palmitoyl Transferase
Gene Aliases
CELE_Y46G5A.17
Chromosome Location
Chr.II: 12788244 - 12796452 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_064320.6 NP_496721.2 2 - 3 408 97

View Full Product Details