Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
bcmo-1
Entrez Gene ID
189959
Gene Name
Beta-Carotene 15,15'-MonoOxygenase
Gene Aliases
CELE_Y46G5A.24
Chromosome Location
Chr.II: 12830676 - 12832717 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_064328.5 NP_496729.2 2 - 3 1078 97

View Full Product Details