Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
gsy-1
Entrez Gene ID
174924
Gene Name
Glycogen [starch] synthase
Gene Aliases
CELE_Y46G5A.31
Chromosome Location
Chr.II: 12866446 - 12878121 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_064335.6 NP_496736.1 6 - 7 1530 64

View Full Product Details