Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
clec-66
Entrez Gene ID
174929
Gene Name
C-type LECtin
Gene Aliases
CELE_F35C5.9
Chromosome Location
Chr.II: 12904398 - 12906686 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_064345.5 NP_496746.1 1 - 2 126 116

View Full Product Details