Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
zyx-1
Entrez Gene ID
174951
Gene Name
protein-coding
Gene Aliases
CELE_F42G4.3
Chromosome Location
Chr.II: 13056814 - 13071530 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001267435.3 NP_001254364.1 5 - 6 1029 69
NM_001267436.3 NP_001254365.1 1 - 2 231 69
NM_064375.8 NP_496776.1 6 - 7 1281 69

View Full Product Details