Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
lec-1
Entrez Gene ID
174964
Gene Name
protein-coding
Gene Aliases
CELE_W09H1.6
Chromosome Location
Chr.II: 13195543 - 13199716 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027234.3 NP_001022405.1 2 - 3 693 72
NM_001383978.1 2 - 3 634 72
GenBank mRNA M94671.1 - 2 - 3 612 72

View Full Product Details