Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
btf-1
Entrez Gene ID
174965
Gene Name
BTAF (TBP-associated factor) homolog
Gene Aliases
CELE_F15D4.1
Chromosome Location
Chr.II: 13200165 - 13215712 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_064401.7 NP_496802.2 9 - 10 2677 58

View Full Product Details