Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
mac-1
Entrez Gene ID
174974
Gene Name
Protein mac-1
Gene Aliases
CELE_Y48C3A.7
Chromosome Location
Chr.II: 13311459 - 13320911 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_064413.8 NP_496814.1 6 - 7 2291 68

View Full Product Details