Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pqn-27
Entrez Gene ID
3564944
Gene Name
protein-coding
Gene Aliases
CELE_E01G4.4
Chromosome Location
Chr.II: 13448835 - 13460642 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001267459.3 NP_001254388.1 5 - 6 784 89
NM_001267460.3 NP_001254389.1 3 - 4 490 89
NM_001383981.1 6 - 7 973 89

View Full Product Details