Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
drr-1
Entrez Gene ID
174994
Gene Name
MARVEL domain-containing protein
Gene Aliases
CELE_F45H10.4
Chromosome Location
Chr.II: 13495046 - 13495900 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_064439.7 NP_496840.1 3 - 4 479 69

View Full Product Details