Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
kqt-3
Entrez Gene ID
191699
Gene Name
protein-coding
Gene Aliases
CELE_Y54G9A.3
Chromosome Location
Chr.II: 13696251 - 13712931 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001267474.4 NP_001254403.1 6 - 7 1283 61
NM_001267475.3 NP_001254404.1 5 - 6 1147 61
NM_001267476.3 NP_001254405.1 5 - 6 1106 61
GenBank mRNA AY572975.1 - 4 - 5 1036 61

View Full Product Details