Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
mcd-1
Entrez Gene ID
175040
Gene Name
protein-coding
Gene Aliases
CELE_Y51H1A.6
Chromosome Location
Chr.II: 13888544 - 13898463 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001267483.3 NP_001254412.1 4 - 5 1534 87

View Full Product Details