Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
sre-44
Entrez Gene ID
189279
Gene Name
Serpentine Receptor, class E (epsilon)
Gene Aliases
CELE_W07G1.2
Chromosome Location
Chr.II: 13949590 - 13951933 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_064517.3 NP_496918.1 1 - 2 291 120

View Full Product Details