Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
eat-2
Entrez Gene ID
175072
Gene Name
Neuronal acetylcholine receptor subunit eat-2
Gene Aliases
CELE_Y48B6A.4
Chromosome Location
Chr.II: 14166878 - 14171658 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_064558.6 NP_496959.1 2 - 3 678 87

View Full Product Details