Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
jmjd-2
Entrez Gene ID
175080
Gene Name
Lysine-specific demethylase 4
Gene Aliases
CELE_Y48B6A.11
Chromosome Location
Chr.II: 14234961 - 14243603 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_064568.5 NP_496969.2 4 - 5 1690 72

View Full Product Details