Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srh-41
Entrez Gene ID
190296
Gene Name
Serpentine Receptor, class H
Gene Aliases
CELE_Y54G11A.12
Chromosome Location
Chr.II: 14285754 - 14287377 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_064573.3 NP_496974.2 1 - 2 334 76

View Full Product Details