Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ctl-2
Entrez Gene ID
175085
Gene Name
Peroxisomal catalase 1
Gene Aliases
CELE_Y54G11A.5
Chromosome Location
Chr.II: 14299540 - 14302027 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027302.7 NP_001022473.1 2 - 3 704 87
GenBank mRNA X82175.1 - - 711 87

View Full Product Details