Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
unc-52
Entrez Gene ID
175126
Gene Name
protein-coding
Gene Aliases
CELE_ZC101.2
Chromosome Location
Chr.II: 14647325 - 14684456 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001267509.3 NP_001254438.1 7 - 8 1200 76
NM_001377905.1 NP_001364664.1 40 - 41 10957 76
NM_064643.10 NP_497044.3 31 - 32 8697 76

View Full Product Details