Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- elo-9
- Entrez Gene ID
-
190207
- Gene Name
- Elongation of very long chain fatty acids protein
- Gene Aliases
-
CELE_Y53F4B.2
- Chromosome Location
-
Chr.II: 14959513 - 14964108 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_064685.2 |
NP_497086.1 |
3 - 4 |
533 |
128 |
View Full Product Details