Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- gst-27
- Entrez Gene ID
-
175166
- Gene Name
- Glutathione S-Transferase
- Gene Aliases
-
CELE_Y53F4B.30
- Chromosome Location
-
Chr.II: 15164890 - 15165731 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_064715.7 |
NP_497116.1 |
1 - 2 |
204 |
117 |
- Gene Symbol
- gst-26
- Entrez Gene ID
-
190223
- Gene Name
- Glutathione S-Transferase
- Gene Aliases
-
CELE_Y53F4B.29
- Chromosome Location
-
Chr.II: 15163808 - 15165716 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_064714.8 |
NP_497115.1 |
1 - 2 |
189 |
117 |
View Full Product Details