Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fbxa-6
Entrez Gene ID
183881
Gene Name
F-box domain-containing protein
Gene Aliases
CELE_cTel54X.1
Chromosome Location
Chr.III: 1219 - 3097 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_064723.2 NP_497124.1 1 - 2 281 75

View Full Product Details