Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
btb-5
Entrez Gene ID
189221
Gene Name
BTB domain-containing protein
Gene Aliases
CELE_W05G11.5
Chromosome Location
Chr.III: 59869 - 61166 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001330882.2 NP_001317757.1 3 - 4 664 115

View Full Product Details