Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ver-1
Entrez Gene ID
175182
Gene Name
Protein ver-1
Gene Aliases
CELE_T17A3.1
Chromosome Location
Chr.III: 156824 - 163542 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_064761.2 NP_497162.2 8 - 9 2629 74
GenBank mRNA AF188751.1 - 8 - 9 2629 74

View Full Product Details