Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fem-2
Entrez Gene ID
175217
Gene Name
Protein phosphatase fem-2
Gene Aliases
CELE_T19C3.8
Chromosome Location
Chr.III: 631941 - 634626 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_064823.9 NP_497224.1 4 - 5 1244 82

View Full Product Details