Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dos-3
Entrez Gene ID
186905
Gene Name
Delta and OSM-11-like
Gene Aliases
CELE_K02F3.8
Chromosome Location
Chr.III: 835751 - 839591 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_064870.8 NP_497271.3 7 - 8 1169 91

View Full Product Details