Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rnp-5
Entrez Gene ID
175244
Gene Name
RRM domain-containing protein
Gene Aliases
CELE_K02F3.11
Chromosome Location
Chr.III: 852799 - 853909 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001384016.1 2 - 3 729 74

View Full Product Details