Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fbxa-21
Entrez Gene ID
191042
Gene Name
FTH domain-containing protein
Gene Aliases
CELE_Y119D3B.9
Chromosome Location
Chr.III: 1235994 - 1237258 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_064951.4 NP_497352.1 1 - 2 111 87

View Full Product Details