Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
fbxa-32
Entrez Gene ID
191060
Gene Name
F-box domain-containing protein
Gene Aliases
CELE_ZC47.6
Chromosome Location
Chr.III: 1267299 - 1269256 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_064962.2 NP_497363.1 2 - 3 721 148

View Full Product Details