Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fbxa-35
Entrez Gene ID
191034
Gene Name
F-box domain-containing protein
Gene Aliases
CELE_Y119D3A.3
Chromosome Location
Chr.III: 1295666 - 1297142 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_064974.3 NP_497375.1 2 - 3 744 155

View Full Product Details