Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fbxa-5
Entrez Gene ID
3565750
Gene Name
F-box domain-containing protein
Gene Aliases
CELE_Y82E9BL.8
Chromosome Location
Chr.III: 1336695 - 1340340 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_064984.4 NP_497385.1 1 - 2 89 76

View Full Product Details