Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- fbxa-5
- Entrez Gene ID
-
3565750
- Gene Name
- F-box domain-containing protein
- Gene Aliases
-
CELE_Y82E9BL.8
- Chromosome Location
-
Chr.III: 1336695 - 1340340 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_064984.4 |
NP_497385.1 |
3 - 4 |
784 |
80 |
- Gene Symbol
- fbxa-26
- Entrez Gene ID
-
190756
- Gene Name
- F-box domain-containing protein
- Gene Aliases
-
CELE_Y82E9BL.7
- Chromosome Location
-
Chr.III: 1336733 - 1342631 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_064985.4 |
NP_497386.2 |
5 - 6 |
2242 |
80 |
View Full Product Details