Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ser-4
Entrez Gene ID
175322
Gene Name
G_PROTEIN_RECEP_F1_2 domain-containing protein
Gene Aliases
CELE_Y22D7AR.13
Chromosome Location
Chr.III: 1723194 - 1733101 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_065051.7 NP_497452.1 1 - 2 165 66
GenBank mRNA U15167.1 - 1 - 2 177 66

View Full Product Details