Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cnc-6
Entrez Gene ID
175324
Gene Name
CaeNaCin (Caenorhabditis bacteriocin)
Gene Aliases
CELE_Y46E12A.1
Chromosome Location
Chr.III: 1763522 - 1764902 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_065054.4 NP_497455.1 1 - 2 145 69

View Full Product Details