Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pqn-82
Entrez Gene ID
175330
Gene Name
Prion-like-(Q/N-rich)-domain-bearing protein
Gene Aliases
CELE_Y39A3CR.7
Chromosome Location
Chr.III: 1849047 - 1854623 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_065063.6 NP_497464.1 1 - 2 82 59

View Full Product Details