Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
fbxa-12
Entrez Gene ID
183326
Gene Name
F-box domain-containing protein
Gene Aliases
CELE_C39B5.7
Chromosome Location
Chr.III: 2219708 - 2222441 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_065112.2 NP_497513.1 2 - 3 754 85

View Full Product Details