Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srd-65
Entrez Gene ID
191823
Gene Name
Serpentine Receptor, class D (delta)
Gene Aliases
CELE_C39B5.12
Chromosome Location
Chr.III: 2222438 - 2224032 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_065113.4 NP_497514.1 1 - 2 221 125

View Full Product Details