Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srd-67
Entrez Gene ID
191825
Gene Name
Serpentine Receptor, class D (delta)
Gene Aliases
CELE_C39B5.11
Chromosome Location
Chr.III: 2244165 - 2245827 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_065120.1 NP_497521.1 5 - 6 789 76

View Full Product Details