Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fbxa-9
Entrez Gene ID
190271
Gene Name
F-box domain-containing protein
Gene Aliases
CELE_Y54F10BM.5
Chromosome Location
Chr.III: 2266651 - 2268315 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_065128.3 NP_497529.2 1 - 2 183 87

View Full Product Details