Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
fbxa-39
Entrez Gene ID
190270
Gene Name
F-box domain-containing protein
Gene Aliases
CELE_Y54F10BM.4
Chromosome Location
Chr.III: 2273032 - 2277219 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001381766.2 NP_001368043.1 3 - 4 860 76

View Full Product Details