Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
apb-1
Entrez Gene ID
175376
Gene Name
AP complex subunit beta
Gene Aliases
CELE_Y71H2B.10
Chromosome Location
Chr.III: 2583953 - 2597034 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027766.4 NP_001022937.1 4 - 5 1803 71

View Full Product Details