Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
twk-45
Entrez Gene ID
190614
Gene Name
protein-coding
Gene Aliases
CELE_Y71H2AM.22
Chromosome Location
Chr.III: 2793323 - 2801900 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001129270.3 NP_001122742.2 9 - 10 1171 70

View Full Product Details