Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cutl-25
Entrez Gene ID
190305
Gene Name
ZP domain-containing protein
Gene Aliases
CELE_Y55D5A.6
Chromosome Location
Chr.III: 3042593 - 3047140 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_065250.3 NP_497651.2 5 - 6 539 67

View Full Product Details