Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
gei-1
Entrez Gene ID
175436
Gene Name
protein-coding
Gene Aliases
CELE_F45H7.2
Chromosome Location
Chr.III: 3344873 - 3363427 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001267939.3 NP_001254868.1 24 - 25 4188 99
NM_001267940.3 NP_001254869.1 2 - 3 572 99
NM_065293.6 NP_497694.3 25 - 26 4419 99
NM_065294.6 NP_497695.3 23 - 24 3892 99

View Full Product Details