Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gei-1
Entrez Gene ID
175436
Gene Name
protein-coding
Gene Aliases
CELE_F45H7.2
Chromosome Location
Chr.III: 3344873 - 3363427 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001267939.3 NP_001254868.1 15 - 16 1588 57
NM_065293.6 NP_497694.3 16 - 17 1819 57
NM_065294.6 NP_497695.3 16 - 17 1820 57

View Full Product Details