Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
prk-2
Entrez Gene ID
175437
Gene Name
Serine/threonine-protein kinase prk-2
Gene Aliases
CELE_F45H7.4
Chromosome Location
Chr.III: 3363666 - 3370209 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_065295.7 NP_497696.2 3 - 4 648 82

View Full Product Details