Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
unc-93
Entrez Gene ID
175466
Gene Name
protein-coding
Gene Aliases
CELE_C46F11.1
Chromosome Location
Chr.III: 3644375 - 3648822 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026047.5 NP_001021218.1 1 - 2 103 71
NM_001381807.1 NP_001366876.1 1 - 2 91 71

View Full Product Details