Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cyp-25A1
Entrez Gene ID
183247
Gene Name
CYtochrome P450 family
Gene Aliases
CELE_C36A4.1
Chromosome Location
Chr.III: 3832591 - 3834586 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_065374.5 NP_497775.3 3 - 4 413 84

View Full Product Details