Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tir-1
Entrez Gene ID
175502
Gene Name
protein-coding
Gene Aliases
CELE_F13B10.1
Chromosome Location
Chr.III: 3869590 - 3901642 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001312937.3 NP_001299866.1 8 - 9 2856 67
NM_001312939.3 NP_001299868.1 2 - 3 942 67
NM_001381812.1 NP_001367074.1 9 - 10 2978 67
NM_065383.4 NP_497784.1 11 - 12 3020 67
GenBank mRNA AY834227.1 - 9 - 10 2858 67

View Full Product Details