Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pat-3
Entrez Gene ID
175504
Gene Name
Integrin beta pat-3
Gene Aliases
CELE_ZK1058.2
Chromosome Location
Chr.III: 3908924 - 3914149 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_065386.6 NP_497787.1 1 - 2 216 79

View Full Product Details